missing translation for 'onlineSavingsMsg'
Learn More

CD99 Antibody (MIC2/8119R), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18588314 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18588314 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18588314

Brand: Novus Biologicals NBP321121AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD99
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon MIC2/8119R
Conjugado Alexa Fluor 750
Formulación 50 mM Sodium Borate
Alias de gen 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Especie del huésped Rabbit
Inmunógeno Recombinant fragment (around aa1-185) of human CD99 protein (exact sequence is proprietary)
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 4267
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.