Learn More
CD97 Mouse anti-Human, Unconjugated, Clone: 3F10C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66972-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The CD97 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD97. This antibody recognizes human, mouse antigen. The CD97 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Especificaciones
| CD97 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Flow Cytometry, ELISA | |
| 3F10C2 | |
| Unconjugated | |
| 150 μL | |
| 976 | |
| IgG2a |