Learn More
CD90 / Thy1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27770-1-AP-20UL
Descripción
The CD90 / Thy1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD90 / Thy1 . This antibody recognizes human, mouse, rat, pig antigen. The CD90 / Thy1 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| CD90 / Thy1 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CD90, CD90 / Thy1 , CDw90, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1 | |
| 20 μL | |
| 7070 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |