missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD90/Thy1 Monoclonal antibody specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | CD90/Thy1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | CL1028 |
| Conjugado | Alexa Fluor 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a recombinant protein derived from P04216, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?