missing translation for 'onlineSavingsMsg'
Learn More

CD83 Antibody (982502), DyLight 755, Novus Biologicals™

Código de producto. 18535756 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18535756 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18535756 Supplier Novus Biologicals Supplier No. FAB17741Z

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD83 Monoclonal antibody specifically detects CD83 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD83
Aplicaciones Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 982502
Conjugado DyLight 755
Dilución Flow Cytometry, CyTOF-ready
Formulación 50mM Sodium Borate
Alias de gen B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83
Especie del huésped Mouse
Inmunógeno Human embryonic kidney cell line HEK293-derived recombinant human CD83, Met1-Ala143, Accession # Q01151
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Dendritic Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 9308
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.