missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD82/Kai-1 Polyclonal specifically detects CD82/Kai-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
| Antígeno | CD82/Kai-1 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulación | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Alias de gen | CD82 antigenC33, CD82 molecule, IA4C33 antigen, Inducible membrane protein R2, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2tetraspanin-27, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN274F9 |
| Símbolos de los genes | CD82 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSICLCRHVHSEDYSKVPKY |
| Mostrar más |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?