missing translation for 'onlineSavingsMsg'
Learn More

CD79A Antibody (JCB117), Novus Biologicals™

Código de producto. 18165432 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mg
0.1 mg
Tamaño de la unidad:
0.02 miligramo
0.10 miligramo
Código de producto. Cantidad unitSize
18165432 0.1 mg 0.10 miligramo
18194374 0.02 mg 0.02 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18165432

Marca: Novus Biologicals NBP2329490.1MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CD79A Monoclonal specifically detects CD79A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD79A
Aplicaciones Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon JCB117
Conjugado Unconjugated
Dilución Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page
Formulación PBS with 0.05% BSA. with 0.05% Sodium Azide
N.º de referencia del gen P11912
Alias de gen CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein
Símbolos de los genes CD79A
Especie del huésped Mouse
Inmunógeno A synthetic peptide corresponding to aa 202-216 (GTYQDVGSLNIADVQ) of human CD79a protein.
Peso molecular del antígeno 44 kDa
Método de purificación Protein A purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Cell Biology, Immunology, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 973
Especificidad de la prueba A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells. This complex of mb-1 and B29 polypeptides and immunoglobulin constitute the B cell Ag receptor. CD79a first appears at pre B cell stage, early in maturation, and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Anti-CD79a is generally used to complement anti-CD20 especially for mature B-cell lymphomas after treatment with Rituximab (anti-CD20). This antibody will stain many of the same lymphomas as anti-CD20, but also is more likely to stain B-lymphoblastic lymphoma/leukemia than is anti-CD20. Anti-CD79a also stains more cases of plasma cell myeloma and occasionally some types of endothelial cells as well.
Especies diana Human
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.