Learn More
CD73 Mouse, anti-Human, PE, Clone: AD2, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60044PE
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17178751
  • 602.10€ / 100 tests
Descripción
  • The AD2 antibody reacts with human CD73, a glycosyl phosphatidylinositol (GPI)-anchored glycoprotein and ecto-5'-nucleotidase expressed on the surface of subsets of B and T cells, follicular dendritic cells, mesenchymal stem cells, endothelial cells, and epithelial cells
  • CD73 catalyzes the hydrolysis of 5'-adenosine monophosphate (AMP) to form the bioactive nucleoside adenosine, and plays a pivotal role in the activation of P1 adenosine receptors by regulating extracellular adenosine concentrations
  • CD73 also appears to function as a co-signaling molecule on T cells and as an adhesion molecule mediating lymphocyte interactions with the endothelium and follicular dendritic cells
  • CD73 is used as a marker for lymphocyte differentiation, its expression increasing during development
  • It is also a useful marker for identifying undifferentiated mesenchymal stem cells
  • CD73 is highly expressed in many types of human and mouse cancers, and has been implicated in the control of tumor growth
Especificaciones
Especificaciones
CD73
Monoclonal
PE
100 Tests
Rhesus Monkey, Chimpanzee, Human, Macaque
Mesenchymal Stem and Progenitor Cells
Immunohistochemistry, Flow Cytometry, Immunofluorescence
AD2
Mouse
Primary
IgG1 κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.