missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | CD7 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CD7/3868R |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Formulación | 10 mM PBS with 0.05% BSA |
| Alias de gen | CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein |
| Especie del huésped | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?