Learn More
CD45RO Antibody (ILA116A), Novus Biologicals™
Código de producto. 18258654 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
0.10 miligramo
18258654 0.1 mg 0.10 miligramo
1 options
Código de producto. 18258654 Proveedor Novus Biologicals N.º de proveedor NBP261035
Mouse Monoclonal Antibody
CD45RO Monoclonal specifically detects CD45RO in Bovine, Goat, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD45RO
Aplicaciones Flow Cytometry, Immunohistochemistry, Functional Assay
Clasificación Monoclonal
Clon ILA116A
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Flow Cytometry 1:10 - 1:1000
Formulación Ascites with 0.09% Sodium Azide
Alias de gen B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Símbolos de los genes PTPRC
Especie del huésped Mouse
Inmunógeno Bovine mononuclear leukocytes
Método de purificación Unpurified
Cantidad 0.1 mg
Disciplina de investigación Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 5788
Especies diana Bovine, Goat, Water Buffalo, Sheep, Feline, Hamster
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG3