Learn More
CD43 Mouse, anti-Human, PE, Clone: CD43-10G7, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 60085PE
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The CD43-10G7 antibody reacts with human CD43, a type I transmembrane sialoglycoprotein expressed on the surface of most human hematopoietic cells, including thymocytes, peripheral T cells, NK cells, monocytes, granulocytes, some B cells (activated B cells and an early B cell subset in bone marrow), plasma cells, and hematopoietic progenitor cells
- Low expression levels are observed on platelets
- CD43 exhibits different glycoforms in the range of ∼95 - 135 kDa, depending on the cell type
- It binds ligands such as CD54 and CD169 (Siglec-1) and plays regulatory roles in cell adhesion and activation, apoptosis and tumor progression, and is used a marker for discriminating normal from neoplastic T cells and B cells within tissues
- Binding of the CD43-10G7 antibody to CD43 on neutrophils reportedly induces homotypic cell adhesion
Especificaciones
| CD43 | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Human | |
| B Cells |
| Immunoprecipitation, Flow Cytometry, Functional Assay | |
| CD43-10G7 | |
| Mouse | |
| Primary | |
| IgG1 κ |