Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Código de producto. 18230960 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.2 mg
Tamaño de la unidad:
0.20 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18230960 0.2 mg 0.20 miligramo
18197689 0.025 mg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18230960 Proveedor Novus Biologicals N.º de proveedor NBP2526580.2MG
Rabbit Monoclonal Antibody
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD4
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Clasificación Monoclonal
Clon YNB46.1.8 (Campath-9H)
Conjugado Unconjugated
Dilución Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready
Formulación PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Alias de gen CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Símbolos de los genes CD4
Especie del huésped Rabbit
Inmunógeno Rat T-cell line (NB2) transfected with the human CD4.
Método de purificación Protein A or G purified
Cantidad 0.2 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 920
Especificidad de la prueba Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG