Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)) - Chimeric - Azide and BSA Free, Novus Biologicals™
Código de producto. 18197689 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.2 mg
Tamaño de la unidad:
0.20 miligramo
25 microgramos
18197689 0.025 mg 25 microgramos
18230960 0.2 mg 0.20 miligramo
2 options
Código de producto. 18197689
Marca: Novus Biologicals NBP2526580.025MG
Rabbit Monoclonal Antibody
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD4
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Clasificación Monoclonal
Clon YNB46.1.8 (Campath-9H)
Conjugado Unconjugated
Dilución Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready
Formulación PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Alias de gen CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Símbolos de los genes CD4
Especie del huésped Rabbit
Inmunógeno Rat T-cell line (NB2) transfected with the human CD4.
Método de purificación Protein A or G purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 920
Especificidad de la prueba Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG