missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD370 (Clec9A) Rat anti-Mouse, BB515, Clone: 10B4, BD Horizon™
Rat Monoclonal Antibody
292.00€
Especificaciones
Antígeno | CD370 (Clec9A) |
---|---|
Clon | 10B4 (also known as 24/04-10B4) |
Concentración | 0.2mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15864699
|
BD Biosciences
566033 |
25 μg |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.Especificaciones
CD370 (Clec9A) | |
0.2mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD370; CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
10B4 (also known as 24/04-10B4) | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
Rat | |
Cell Biology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Clec9A Peptide | |
Primary |
For Research Use Only.