missing translation for 'onlineSavingsMsg'
Learn More

CD36 Antibody (185-1G2), PE, Novus Biologicals™

Código de producto. 18027256 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18027256 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18027256

Marca: Novus Biologicals NBP295271PE

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD36/SR-B3
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 185-1G2
Conjugado PE
Formulación PBS
Alias de gen CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor
Especie del huésped Mouse
Inmunógeno Stimulated human leukocytes
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 948
Especificidad de la prueba Recognizes a protein of 80kDa-90kDa, identified as CD36 (Workshop IV; Code P-26). Its epitope maps between aa155-183. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischemia observed in sickle cell disease and cerebral malaria. Note that 1-4% of Japanese and East Asia population lack CD36. This monoclonal antibody blocks adhesion of P. falciparum parasitized red blood cells to CD36 and strongly inhibits collagen-induced platelet aggregation.
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.