missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting.
Especificaciones
Especificaciones
| Antígeno | CD36/SR-B3 |
| Aplicaciones | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 650 |
| Dilución | Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| N.º de referencia del gen | P16671 |
| Alias de gen | CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor |
| Símbolos de los genes | CD36 |
| Especie del huésped | Rabbit |
| Mostrar más |
For Research Use Only
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?