Learn More
CD36 Antibody - BSA Free, Novus Biologicals™
Código de producto. p-5062298 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 ml
25 microlitros
Cantidad:
0.1 mL
Código de producto. unitSize Cantidad
18756803 25 microlitros -
18765263 0.10 ml 0.1 mL
2 options
Código de producto. 18756803
Marca: Novus Biologicals NB400145SS
Rabbit Polyclonal Antibody has been used in 9 publications
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, SDS-Page.
TRUSTED_SUSTAINABILITY
Especificaciones
Inmunógeno A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProt# P16671]