missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Marca: Novus Biologicals NB400-145
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, SDS-Page.
Especificaciones
| CD36/SR-B3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Rabbit | |
| 75 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Human, Mouse, Amphibian | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
| 1 mg/mL | |
| Western Blot 1:2000-1:10000, Simple Western 1:40, Flow Cytometry reported in scientific literature (PMID 32971872), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27226602), Immunoblotting reported in scientific literature (PMID 31501473), SDS-Page reported in scientific literature (PMID 31501473) | |
| P16671 | |
| CD36 | |
| A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProt# P16671] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human.Mouse reactivity reported in scientific literature (PMID: 24163073) Immunogen sequence has 95% homology to rat, monkey, Golden hamster and rabbit, 90% homology to mouse, and 75% homology to bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido