missing translation for 'onlineSavingsMsg'
Learn More

CD35 Antibody (8C12), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™

Código de producto. 18756421 Tienda Bio Techne Productos
Change view
Click to view available options
:
0.1 mL, Alexa Fluor 594
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto.
18756421 0.1 mL, Alexa Fluor 594
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18756421 Supplier Novus Biologicals Supplier No. NBP252668AF594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD35
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Clasificación Monoclonal
Clon 8C12
Conjugado Alexa Fluor 594
Formulación 50 mM Sodium Borate
Alias de gen complement component (3b/4b) receptor 1 (Knops blood group), KN
Especie del huésped Rabbit
Inmunógeno Purified murine CR1 protein.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 1378
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.