missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD34 Monoclonal antibody specifically detects CD34 in Human, Primate, Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
| Aplicaciones | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Especies diana | Bovine (Negative), Canine (Negative), Rat (Negative) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?