Learn More
CD31/PECAM-1 Antibody (MEC13.3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 17 publications
Marca: Novus Biologicals NB600-1475
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.
Especificaciones
| CD31/PECAM-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:100, In vitro assay reported in scientific literature (PMID 24647208), In vivo assay reported in scientific literature, Flow (Cell Surface) 2.5 ug/ml, CyTOF-ready | |
| PECAM1 | |
| This CD31/PECAM-1 Antibody (MEC13.3) was developed against mouse endothelial cell line T-end. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, CyTOF | |
| MEC13.3 | |
| Unconjugated | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, MEC 13.3 CD31, MEC 13.3 Clone, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Rat | |
| 82.5 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 5175 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido