Learn More
CD31/PECAM-1 Antibody (CL14693) [mFluor Violet 500 SE], Novus Biologicals™
Código de producto. 30301045 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30301045 0.1 mL 0.10 ml
1 options
Código de producto. 30301045
Marca: Novus Biologicals NBP326877MFV500
Mouse Monoclonal Antibody
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD31/PECAM-1
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon CL14693
Conjugado mFluor Violet 500 SE
Formulación 50mM Sodium Borate
Alias de gen adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from P16284, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 5175
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1