missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | CD31/PECAM-1 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | C31/9224R |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Formulación | 10 mM PBS with 0.05% BSA |
| Alias de gen | adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule |
| Especie del huésped | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?