missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300f/LMIR3/CD300LF Antibody (234903), CoraFluor™ 1, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
CD300f/LMIR3/CD300LF Monoclonal antibody specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Especificaciones
Especificaciones
| Antígeno | CD300f/LMIR3/CD300LF |
| Aplicaciones | Flow Cytometry, Immunocytochemistry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 234903 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CLM1, CLM-1, IgSF13, IREM1, IREM-1, NKIR |
| Especie del huésped | Rat |
| Inmunógeno | Y3 rat myeloid cell line transfected with human CD300f/LMIR3 |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?