missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , PE, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566683
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Especificaciones
| CD3 | |
| Monoclonal | |
| PE | |
| CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
| Sheep Erythrocyte Rosette-purified Human T Cells | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| OKT3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido