missing translation for 'onlineSavingsMsg'
Learn More

CD3 epsilon Antibody (TR66), DyLight 488, Novus Biologicals™

Código de producto. 18125610 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18125610 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. View return policy

Product Code. 18125610

Brand: Novus Biologicals NBP197446G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD3 epsilon
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon TR66
Conjugado DyLight 488
Dilución Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Símbolos de los genes CD3E
Especie del huésped Mouse
Inmunógeno Cloned human helper T cell line.
Método de purificación Protein G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 916
Especificidad de la prueba Recognizes the epsilon-chain of human CD3.
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.