missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for ELISA.
Especificaciones
Especificaciones
| Antígeno | CD27 Ligand/TNFSF7/CD70 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | TAN 1-7 |
| Conjugado | HRP |
| Dilución | ELISA |
| Formulación | PBS with No Preservative |
| Alias de gen | CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 |
| Símbolos de los genes | CD70 |
| Especie del huésped | Rat |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?