Learn More
CD2 Mouse, anti-Human, PE, Clone: RPA-2.10, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca:  Stemcell Technologies 60007PE
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17178381
  • 464.40€ / 100 tests
Descripción
  • The RPA-2.10 antibody reacts with CD2, an ∼50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion
  • CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium
  • CD2 expression appears early during T cell differentiation
  • Aberrant expression has been observed in some lymphomas and myeloid leukemias
  • CD2 is critically important for T cell activation and signaling, as well as lymphocyte adhesion
  • The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59
  • Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction
Especificaciones
Especificaciones
CD2
Monoclonal
PE
100 Tests
Baboon, Cynomolgus, Pig, Human
T Cells
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
RPA-2.10
Mouse
Primary
IgG1 κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.