missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD2/CD7 Antibody Cocktail, FITC, PE

Antibody Cocktail
345.00€ - 564.00€
Specifications
| Antígeno | CD2/CD7 |
|---|---|
| Clon | T6.3, MG34, MG34 |
| Contenido y almacenamiento | 4°C, store in dark, DO NOT FREEZE! |
| Aplicaciones | Flow Cytometry |
| Clasificación | Cocktail |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
11513483
![]() |
Invitrogen™
MA112004 |
100 Tests |
564.00€
100 tests |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
11523483
|
Invitrogen™
MA112005 |
50 Tests |
345.00€
50 tests |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD7 (clone MG34, isotype IgG2a).
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Specifications
| CD2/CD7 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cocktail | |
| Liquid | |
| IgG2a | |
| Human | |
| CD7 | |
| 2CD2 antigen; CD2; CD2 molecule; CD7; CD7 antigen (p41); CD7 molecule; Erythrocyte receptor; GP40; LEU-9; LFA-2; LFA-3 receptor; p41 protein; Rosette receptor; SRBC; T11; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen CD2; T-cell surface antigen Leu-9; T-cell surface antigen T11/Leu-5; Tp40; TP41 | |
| CD2, CD7 | |
| Primary | |
| Protein A/G |
| T6.3, MG34, MG34 | |
| Flow Cytometry | |
| FITC/PE | |
| Mouse | |
| RUO | |
| PBS with 0.2% carrier protein and 0.08% sodium azide | |
| P06729, P09564 | |
| 914, 924 | |
| T lymphocytes activated by mixed lymphocyte culture and T-acute lymphoblastic leukemia (T-ALL) cells. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title