missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD2/CD7 Antibody Cocktail, FITC, PE

Código de producto. 11523483
Change view
Click to view available options
Cantidad:
100 Tests
50 Tests
Tamaño de la unidad:
100 tests
50 tests
Código de producto. Cantidad unitSize
11523483 50 Tests 50 tests
11513483 100 Tests 100 tests
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11523483

Marca: Invitrogen™ MA112005

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD2 (clone T6.3, isotype IgG2a) and PE-conjugated CD7 (clone MG34, isotype IgG2a).

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD2/CD7
Aplicaciones Flow Cytometry
Clasificación Cocktail
Clon T6.3, MG34, MG34
Conjugado FITC/PE
Formulación PBS with 0.2% carrier protein and 0.08% sodium azide
génica CD7
N.º de referencia del gen P06729, P09564
Alias de gen 2CD2 antigen; CD2; CD2 molecule; CD7; CD7 antigen (p41); CD7 molecule; Erythrocyte receptor; GP40; LEU-9; LFA-2; LFA-3 receptor; p41 protein; Rosette receptor; SRBC; T11; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen CD2; T-cell surface antigen Leu-9; T-cell surface antigen T11/Leu-5; Tp40; TP41
Símbolos de los genes CD2, CD7
Especie del huésped Mouse
Inmunógeno T lymphocytes activated by mixed lymphocyte culture and T-acute lymphoblastic leukemia (T-ALL) cells.
Método de purificación Protein A/G
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 914, 924
Especies diana Human
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.