missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CD1d Monoclonal specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | CD1d |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1B1 |
| Concentración | 0.2 mg/mL |
| Conjugado | Biotin |
| Dilución | Flow Cytometry 1:10-1:1000 |
| Formulación | Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide |
| Alias de gen | antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D |
| Símbolos de los genes | CD1D |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?