Learn More
CD16/CD32 Rat, anti-Mouse, FITC, Clone: 2.4G2, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60161FI
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17129221
  • 226.80€ / 100 microgramos
Descripción
  • The 2.4G2 antibody reacts with a shared epitope of Fc-gamma receptors CD16 (FcRIII) and CD32 (FcRII) in the mouse
  • CD16 and CD32 are low affinity receptors for the Fc portion of IgG, with CD32 having a higher affinity for IgG aggregates and immune complexes
  • CD16 and CD32 are expressed on B cells, monocytes, macrophages, NK cells, neutrophils, granulocytes, mast cells, and dendritic cells
  • The binding of immunoglobulins to Fc receptors is involved in multiple immunological functions, including phagocytosis, cytotoxocity, and the modulation of immune responses
  • The 2.4G2 antibody blocks the interaction between IgG and CD16 or CD32, and therefore is used as a pre-treatment to minimize non-specific binding of antibodies to FcR-bearing cells in applications such as flow cytometry, cell sorting, and immunofluorescence
Especificaciones
Especificaciones
CD16/CD32
Monoclonal
FITC
100 μg
Mouse
Dendritic Cells, Granulocytes and Subsets, Macrophages, Monocytes, Myeloid Cells, NK Cells, T Cells, T Cells, Other Subsets
Immunoprecipitation, Flow Cytometry, Immunofluorescence, Functional Assay
2.4G2
Rat
Primary
IgG2b κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.