missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138/Syndecan-1 Mouse anti-Human, Clone: 4H5H5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60185-2-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CD138 / Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.Especificaciones
CD138/Syndecan-1 | |
Monoclonal | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18827 | |
SDC1 | |
CD138/Syndecan-1 Fusion Protein Ag0823 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
4H5H5 | |
Unconjugated | |
SDC1 | |
CD138, SDC, SDC1, SYND1, syndecan, syndecan 1 | |
Mouse | |
Protein A | |
RUO | |
6382 | |
-20°C | |
Liquid |