missing translation for 'onlineSavingsMsg'
Learn More

CD11c Antibody (118/A5), DyLight 405, Novus Biologicals™

Product Code. 18722982 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18722982 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722982 Supplier Novus Biologicals Supplier No. NBP250130V

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD11c
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 118/A5
Conjugado DyLight 405
Formulación 50mM Sodium Borate
Alias de gen 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Especie del huésped Mouse
Inmunógeno Synthetic peptide conjugated to KLH corresponding to the C terminus of human CD11C: ANGQIAPENGTQTPSPPSEK
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Dendritic Cell Markers, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 3687
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.