missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD105 (Endoglin) Mouse anti-Human, BB700, Clone: 266, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566528
Detalles adicionales : Peso : 0.00840kg
Descripción
CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow Cytometry.Especificaciones
CD105 (Endoglin) | |
Monoclonal | |
BB700 | |
100 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
266 | |
Mouse | |
RUO | |
CD105 (Endoglin) | |
Refrigerate (2 to 8°C) |