Learn More
Novus Biologicals™ CCR5 Antibody (leronlimab) - Humanized, Novus Biologicals™
Código de producto. 30044624 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
30044624 100 μg 100 microgramos
30047474 50 μg 50 microgramos
30048460 1 mg 1 miligramo
3 options
Código de producto. 30044624
Marca: Novus Biologicals™ NBP328784100ug
Human Monoclonal Antibody
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CCR5
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon leronlimab
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen P51681
Alias de gen C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5
Especie del huésped Human
Inmunógeno CCR5/ CD195
Método de purificación Protein A purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 1234
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG4