missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC72 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20393-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CCDC72, also named as TMA7, is a novel modulator for hair follicle development. CCDC72 has the promoter activity and regulation of gene expression.Especificaciones
CCDC72 | |
Polyclonal | |
Unconjugated | |
TMA7 | |
CCDC72, HSPC016, HSPC330, TMA7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51372, 66167, 679747 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K003, Q9Y2S6 | |
TMA7 | |
CCDC72 Fusion Protein Ag13792 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |