missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx2 Antibody (RAB-C238), Alexa Fluor™ 488 - Chimeric, Novus Biologicals™
Tienda Bio Techne ProductosDescription
Cbx2 Monoclonal antibody specifically detects Cbx2 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antígeno | Cbx2 |
| Aplicaciones | ELISA, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | RAB-C238 |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX2 protein under non-denaturing conditions. |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?