missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NB100-56181
This item is not returnable.
View return policy
Description
CARD8 Polyclonal specifically detects CARD8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| CARD8 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Apoptotic protein NDPP1, CARD inhibitor of NF-kappaB-activating ligands, CARDINALFLJ18121, CARD-inhibitor of NF-kappa-B-activating ligand, caspase recruitment domain family, member 8, caspase recruitment domain-containing protein 8, DACAR, Dakar, KIAA0955DKFZp779L0366, NDPP, NDPP1MGC57162, TUCANFLJ18119, Tumor up-regulated CARD-containing antagonist of CASP9, tumor up-regulated CARD-containing antagonist of caspase nine | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| The gene encoding TUCAN produces, by alternative splicing, multiple transcripts, with introns, putatively encoding numerous different protein isoforms. Thus the molecular weight observed by SDS-PAGE can vary depending on the TUCAN form expressed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| CARD8 | |
| Full-length recombinant human TUCAN (CARD8) protein was used as immunogen. The recombinant protein sequence corresponds to GenBank no. gi 14424229 sp Q9Y2G2 CARD8_HUMAN. | |
| 0.05 mL | |
| Apoptosis | |
| 22900 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction