missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1/CPA1 Antibody (CL6607) [Alexa Fluor™ 488], Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Carboxypeptidase A1/CPA1 Monoclonal antibody specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Carboxypeptidase A1/CPA1 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL6607 |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, EC 3.4.17, EC 3.4.17.1 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a recombinant protein derived from P15085, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?