Learn More
Carbonic Anhydrase IX/CA9 Antibody (66.4.C2 (PN-15)), Novus Biologicals™
Código de producto. 18189922 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
0.2 mg
20 ug
Tamaño de la unidad:
0.10 miligramo
0.20 miligramo
20 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18189922 0.1 mg 0.10 miligramo
18114382 0.2 mg 0.20 miligramo
18472112 20 ug 20 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18189922 Proveedor Novus Biologicals N.º de proveedor NBP2151990.1MG
Mouse Monoclonal Antibody has been used in 5 publications
Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Renal Cell Carcinoma (gp200)
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 66.4.C2 (PN-15)
Concentración 0.2mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Formulación PBS with 0.05% BSA. with 0.05% Sodium Azide
N.º de referencia del gen Q16790
Alias de gen CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Símbolos de los genes LY75
Especie del huésped Mouse
Inmunógeno Fresh, normal human renal cortical tissue homogenate was used as immunogen to generate the RCC antibody.
Método de purificación Protein G purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 4065
Especificidad de la prueba Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Its epitope resides in the carbohydrate domain of gp200. It shows no significant cross-reactivity with other carbohydrate determinants, such as the Lewis blood group antigens, epithelial membrane antigen, HMFG, and AB blood group antigens. In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas. This MAb may be useful in the investigations of carcinomas of proximal nephrogenic differentiation especially those showing tubular differentiation.
Especies diana Human, Equine
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG2b κ
Mostrar más Mostrar menos
For Research Use Only