missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capicua Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB110-59905
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot (Negative).
Especificaciones
| Capicua | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
| Rabbit | |
| 164 kDa | |
| 0.1 mg | |
| Cancer | |
| 23152 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
| Q96RK0 | |
| CIC | |
| The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG |
Corrección del contenido de un producto
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction