missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPG Mouse anti-Human, Clone: 3A5B2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66277-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in non-muscle cells. Alternatively spliced transcript variants have been observed for this gene.Especificaciones
| CAPG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P40121 | |
| CAPG | |
| CAPG Fusion Protein Ag24676 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3A5B2 | |
| Unconjugated | |
| CAPG | |
| Actin regulatory protein CAP G, AFCP, CAPG, Macrophage capping protein, MCP | |
| Mouse | |
| Protein G | |
| RUO | |
| 822 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido