missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAMK4 (CaMKIV), GST Tag Recombinant Protein

Código de producto. 11593417
Change view
Click to view available options
Cantidad:
10 μg
1 mg
Tamaño de la unidad:
1 miligramo
10 microgramos
Código de producto. Cantidad unitSize
11593417 10 μg 10 microgramos
10379172 1 mg 1 miligramo
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11593417

Marca: Invitrogen™ PV3310

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

CAMK4 (CaMKIV) is a serine/threonine protein kinase that phosphoryates CREB, oncoprotein 18, and SRF. It has limited tissue distribution, and has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells. The nuclear localization of this protein is consistent with its role in mediating calcium-dependent gene expression. CaMKIV is particularly abundant in testis, T-cells, and neurons but is also found in other tissues to varying degrees. In neurons, CaMKIV is thought to play an important role in synaptic plasticity via its gene regulatory effects. In T-cells, this protein plays an important role in calcium signaling which could affect the transcription regulatory protein, nuclear factor of activated T-cells (NFAT). CaMKIV is encoded, along with calspermin, by the CaMKIV gene. It has been found that, in testes, CaMKIV is expressed in germ cells and found to be associated with chromatin. The association of CaMKIV with chromatin suggests a potential role in chromatin remodeling during nuclear condensation in spermatids.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q16566
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris with 50% glycerol, 0.05% Triton X-100, 2mM DTT, 150mM NaCl, 0.05mM EDTA and no preservative; pH 7.5
ID de gen (Entrez) 814
Peso molecular 79.6 kDa
Nombre Human CAMK4 (CaMKIV), GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen A430110E23Rik; AI666733; brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Ca2+/calmodulin-dependent protein kinase type IV/Gr; Calcium/calmoduli; calcium/calmodulin dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV; calcium/calmodulin-dependent protein kinase IV L homeolog; calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calmodulin-dependent protein kinase IV; calspermin; CAM kinase- GR; CAM kinase IV; CAM kinase-GR; CAMK; CaMK IV; camk4; camk4.L; camk4-A; CAMK-GR; CAMKIV; CaMKIV/Gr; Ccdpk; D18Bwg0362e; EC 2.7.11.17; KCC4; kinase CaMK4; OTTHUMP00000222818; RATCCDPK; xCaM; XELAEV_18008011mg
Nombre común CaMKIV
Símbolo de gen CAMK4
Familia Quinasa CAMK1 Kinase Family
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Grupo de cinasas Ser⁄Thr Kinases: CAMK Group
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™
Entrada diana CAMK4
Gen diana calcium⁄calmodulin-dependent protein kinase IV
Sistema de expresión E. coli
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Full Length
Formulario Liquid
Subtipo de proteína Serine/Threonine Kinases
Categoría de investigación Signal Transduction
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.