missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CAMK2A (CaMKII Alpha), His Tag Recombinant Protein

Código de producto. 10217492
Change view
Click to view available options
Cantidad:
10 μg
1 mg
Tamaño de la unidad:
1 miligramo
10 microgramos
Código de producto. Cantidad unitSize
10217492 1 mg 1 miligramo
11539326 10 μg 10 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10217492

Marca: Invitrogen™ PR4586A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

CAMK2A (CaMKII alpha) is a serine/threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.
TRUSTED_SUSTAINABILITY

Spezifikation

Número de acceso Q9UQM7
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 20mM tris with 50% glycerol, 100mM NaCl, 0.5mM EDTA, 2mM DTT, 0.05% NP-40 and no preservative; pH 7.5
ID de gen (Entrez) 815
Peso molecular 56.8 kDa
Nombre Human CAMK2A (CaMKII Alpha), His Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 1 mg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; caM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; CaM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975
Nombre común CaMKII alpha
Símbolo de gen CAMK2A
Familia Quinasa CAMK2 Kinase Family
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Grupo de cinasas Ser⁄Thr Kinases: CAMK Group
Condiciones de envío Dry Ice
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Full Length
Formulario Liquid
Subtipo de proteína Serine/Threonine Kinases
Categoría de investigación Signal Transduction
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.