Learn More
Calretinin Antibody (CL12842) [DyLight 755], Novus Biologicals™
Código de producto. 30288731 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30288731 0.1 mL 0.10 ml
1 options
Código de producto. 30288731
Marca: Novus Biologicals NBP318545IR
Mouse Monoclonal Antibody
Calretinin Monoclonal antibody specifically detects Calretinin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antígeno Calretinin
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL12842
Conjugado DyLight 755
Formulación 50mM Sodium Borate
Alias de gen 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P22676, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cellular Markers, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 794
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1