missing translation for 'onlineSavingsMsg'
Learn More

Caldesmon/CALD1 Antibody, Novus Biologicals™

Código de producto. 18227936 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Unit Size:
0.10 ml
25 microlitros
Product Code. Cantidad unitSize
18227936 0.1 mL 0.10 ml
18455711 25 μL 25 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18227936

Brand: Novus Biologicals NBP185702

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 4 publications

Caldesmon/CALD1 Polyclonal specifically detects Caldesmon/CALD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Caldesmon/CALD1
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Símbolos de los genes CALD1
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:SLKIEERAEFLNKSVQKSSGVKSTHQAAIVSKIDSRLEQYTSAIEGTKSAKPTKPAASDLPVPAEGVRNIKSMWEKGNVFSSPTAAGTPNKETAGLKVGVSSRINEWLTKTPDGNKSPAPKPSDLRPGDVSSKRNLWEKQSVDKVTS
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cytoskeleton Markers
Primario o secundario Primary
ID de gen (Entrez) 800
Especificidad de la prueba Specificity of human Caldesmon/CALD1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.