Learn More
Novus Biologicals™ Cadherin-17 Antibody (PTA001-A4) - Humanized, Novus Biologicals™
Código de producto. 30048691 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30048691 50 μg 50 microgramos
30048534 100 μg 100 microgramos
30044101 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30048691 Proveedor Novus Biologicals™ N.º de proveedor NBP32882050ug
Human Monoclonal Antibody
Cadherin-17 Monoclonal antibody specifically detects Cadherin-17 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Cadherin-17
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon PTA001-A4
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q12864
Alias de gen cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024
Especie del huésped Human
Inmunógeno CDH17/ Cadherin-17
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cancer
Primario o secundario Primary
ID de gen (Entrez) 1015
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1