Learn More
Novus Biologicals™ CACNA1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Novus Biologicals™ NBP3-30953-100ug
 Ver más versiones de este producto
Código de producto. 30067031
  • 508.00€ / 100 microgramos
Descripción
CACNA1A Polyclonal antibody specifically detects CACNA1A in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CACNA1A
Polyclonal
Western Blot : 1:400, Immunohistochemistry
APCA, BI, brain calcium channel 1, Brain calcium channel I, CACH4, CACN3, CACNL1A4CAV2.1, calcium channel, L type, alpha-1 polypeptide, Calcium channel, L type, alpha-1 polypeptide isoform 4, calcium channel, voltage-dependent, P/Q type, alpha 1A subunit, Cav2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6, voltage-dependent P/Q-type calcium channel subunit alpha-1A, Voltage-gated calcium channel subunit alpha Cav2.1
Synthetic peptide derived from the rat CACNA1A conjugated to KLH
100 μg
Signal Transduction
773
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunohistochemistry, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified