missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF5/CTRP5 Antibody (332923), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 30062349 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30062349 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30062349

Brand: Novus Biologicals FAB3167S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C1qTNF5/CTRP5 Monoclonal antibody specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno C1qTNF5/CTRP5
Aplicaciones Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 332923
Conjugado Alexa Fluor 750
Dilución Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulación 50mM Sodium Borate
Alias de gen C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3
Especie del huésped Mouse
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human CTRP5/C1qTNF5, Ser16-Ala243 (Gln44Arg), Accession # Q9BXJ0
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Vision
Primario o secundario Primary
ID de gen (Entrez) 114902
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.